Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ STP 120 Spin Tissue Processor
GSA_VA

Catalog No. 22050853 Shop All Epredia Products
Change view
Click to view available options
Includes:
Vent.System, 2 baths, 1 basket
Vent.System, 3 baths, 2 basket
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-050-853 Vent.System, 2 baths, 1 basket
22-050-854 Vent.System, 3 baths, 2 basket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-050-853 Supplier Epredia™ Supplier No. 813150
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Process biological specimens from chemical fixation to paraffin infiltration with the Epredia™ STP 120 Spin Tissue Processor.

  • Immediate and delayed start processing modes
  • Programmable spinning speed of either 60 or 70 rpm; spinning can also be programmed off
  • Programmable immersion time in each station (from 1 minute to 99 hours: 59 minutes)
  • Basket capacity of 120 or 240 cassettes
  • Battery backup system in case of power failure

Performance:

  • Reagent vessel tops and charcoal-enhanced ventilation help control processing vapors
  • Up to 10 different processing programs maintained
  • Cassette baskets spin counterclockwise and clockwise within reagent container to improve processing
  • Reagent carryover is reduced through gentle centrifugal spinning of basket above reagent vessel
  • Each reagent vessel holds 1.8 L of processing reagent

Physical Specifications:

  • Electrical Requirements: 100-240 V, 50/60 Hz
  • Diameter: 85 cm (33.5 in.)
  • Height: 50 cm opening to 70 cm (19.6 in. opening to 27.5 in.)
  • Weight: 154 lb. (70 kg)

Includes:

  • Basic instrument
  • Instruction manual
  • Ten reagent vessels and two paraffin baths for single basket operation (120-cassette capacity) or nine reagent vessels and three paraffin baths for double basket operation (240-cassette capacity)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Basket Capacity 120 or 240 Cassettes
Reagent Capacity 1.8 L
Includes Vent.System, 2 baths, 1 basket
Diameter (English) 33.5 in.
Diameter (Metric) 85 cm
Height (English) 19.6 to 27.5 in.
Height (Metric) 50 to 70 cm
Weight (English) 154 lb.
Weight (Metric) 70 kg
Frequency 50/60 Hz
Voltage 100/240 V
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.