Learn More
Saf T Pak Inc STP-300 - Category A Frozen Insulated Shipping System (Overpack for UN 2900, UN 2814), 4/Case
SDP
Catalog No. NC9238882 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
STP-300 Overpack, Category A Shipping; Insulated; For use with Saf-T-Temp products or Dry ice for thermal control; Outer box O.D.: 15.5 x 13.75 x 12.75in.; Insulated chest I.D.: 12 x 10.25 x 8in.
Catalog No. NC9238882 Supplier Saf T Pak Inc Supplier No. STP300
STP-300 Overpack, Category A Shipping; Insulated; For use with Saf-T-Temp products or Dry ice for thermal control; Outer box O.D.: 15.5 x 13.75 x 12.75in.; Insulated chest I.D.: 12 x 10.25 x 8in.