Learn More
Saf T Pak Inc STP-340 - Category B, Exempt Insulated Shipping System (UN 3373), Frozen with Disposable Secondary Pressure Vessel
SDP
Catalog No. NC9059389 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
STP-340 Category B / Exempt Insulated Shipping System, with Disposable Secondary Pressure Vessel, Large insulated dry ice shipper designed specifically for Category B / Exempt Specimens. 2/Carton
Catalog No. NC9059389 Supplier Saf T Pak Inc Supplier No. STP340
STP-340 Category B / Exempt Insulated Shipping System, with Disposable Secondary Pressure Vessel, Large insulated dry ice shipper designed specifically for Category B / Exempt Specimens. 2/Carton