Learn More
Saf T Pak Inc STP-309DI - CATEGORY B FROZEN INSULATED SHIPPING SYSTEM, (UN3373)
SDP
Catalog No. NC0404148 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
STP-309DI Insulated Category B / Exempt Shipping System, Outer Box – OD 11.25 x 9.5 x 8.5in., pre-printed with Biological Substance, Category B and UN 3373 Markings Inner Box - OD 7.75 x 4.25 x 2.25in., Insulated Chest – ID 7.75 x 5.75 x 5in. 8/case
Catalog No. NC0404148 Supplier Saf T Pak Inc Supplier No. STP309DI
STP-309DI Insulated Category B / Exempt Shipping System, Outer Box – OD 11.25 x 9.5 x 8.5in., pre-printed with Biological Substance, Category B and UN 3373 Markings Inner Box - OD 7.75 x 4.25 x 2.25in., Insulated Chest – ID 7.75 x 5.75 x 5in. 8/case