Learn More
Chemglass Life Sciences Stopcock, 90°, Hollow Glass Plug, High Vacuum, 4mm Bore, 17.35/50 Plug Size, 10mm OD Sidearms
SDP
Catalog No. CG47802
Encompass_Preferred
Similar to CG-476, but with the sidearms sealed at a 90 degrees angle to each other. Precision ground and hand-lapped with corresponding numbers on barrel and plug.
Catalog No. CG47802 Supplier Chemglass Life Sciences Supplier No. CG47802
Similar to CG-476, but with the sidearms sealed at a 90 degrees angle to each other. Precision ground and hand-lapped with corresponding numbers on barrel and plug.