Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Hollow Glass Plug, High Vacuum, 2mm Bore, 12/30 Plug Size, 8mm OD Sidearms, #1 Clip Size
SDP
Catalog No. CG47101
Encompass_Preferred
Hollow plug, straight bore stopcock is precision ground and hand-lapped with corresponding numbers on barrel and plug. They are supplied complete with the listed size CG-423 metal retaining clip.
Catalog No. CG47101 Supplier Chemglass Life Sciences Supplier No. CG47101
Hollow plug, straight bore stopcock is precision ground and hand-lapped with corresponding numbers on barrel and plug. They are supplied complete with the listed size CG-423 metal retaining clip.