Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Hollow Glass Plug, High Vacuum, 4mm Bore, 17/40 Plug Size, 10mm OD Sidearms, #4 Clip Size
SDP
Catalog No. CG47102
Encompass_Preferred
Hollow plug, straight bore stopcock is precision ground and hand-lapped with corresponding numbers on barrel and plug. They are supplied complete with the listed size CG-423 metal retaining clip.
Catalog No. CG47102 Supplier Chemglass Life Sciences Supplier No. CG47102
Hollow plug, straight bore stopcock is precision ground and hand-lapped with corresponding numbers on barrel and plug. They are supplied complete with the listed size CG-423 metal retaining clip.