Learn More
Chemglass Life Sciences Stopcock, Single Oblique, Glass Plug, High Vacuum, 4mm Bore, 17.35/50 Plug Size, 10mm OD Sidearms
SDP
Catalog No. 50873736
Encompass_Preferred
Solid plug, oblique bore stopcock is precision ground with corresponding numbers on barrel and plug. Lower vacuum chamber is evacuated with a separate hole in the plug which is not in the line of flow.
Catalog No. 50-873-736 Supplier Chemglass Life Sciences Supplier No. CG47302
Solid plug, oblique bore stopcock is precision ground with corresponding numbers on barrel and plug. Lower vacuum chamber is evacuated with a separate hole in the plug which is not in the line of flow.