Learn More
Chemglass Life Sciences Stopcock, T-Bore, Glass Plug, High Vacuum, 2mm Bore, 12/30 Plug Size, 8mm OD Sidearms
SDP
Catalog No. CG46401
Encompass_Preferred
Solid glass high vacuum stopcock with theChemglass improved constant pressure retention device. This retaining system incorporates a glass plug having an extended snelling which accepts a stainless steel spring and clip. The spring keeps constant tension on the plug at all times and is easily removed and attached. Similar to CG-462, but with three stems sealed 90 degrees apart. Solid glass plug has a "T" shaped bore so that any two or all three arms can be connected at one time. Plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug. Each stopcock is supplied complete with a stainless steel spring and clip. Replacement retention kit consists of one spring and clip for the plug size listed.
Catalog No. CG46401 Supplier Chemglass Life Sciences Supplier No. CG46401
Solid glass high vacuum stopcock with theChemglass improved constant pressure retention device. This retaining system incorporates a glass plug having an extended snelling which accepts a stainless steel spring and clip. The spring keeps constant tension on the plug at all times and is easily removed and attached. Similar to CG-462, but with three stems sealed 90 degrees apart. Solid glass plug has a "T" shaped bore so that any two or all three arms can be connected at one time. Plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug. Each stopcock is supplied complete with a stainless steel spring and clip. Replacement retention kit consists of one spring and clip for the plug size listed.