Learn More
Chemglass Life Sciences Stopcock, Double Oblique, Glass Plug, Pressure, 2mm Bore, 14.5/50 Plug Size, 8mm OD Sidearms
SDP
Catalog No. CG46001
Encompass_Preferred
Similar to CG-458, but with two parallel oblique bores. Stopcock has a precision ground barrel and plug.
Catalog No. CG46001 Supplier Chemglass Life Sciences Supplier No. CG46001
Similar to CG-458, but with two parallel oblique bores. Stopcock has a precision ground barrel and plug.