Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Glass Plug, Pressure, 2mm Bore, 12/30 Plug Size, 8mm OD Sidearms
SDP
Catalog No. 501531753
Encompass_Preferred
Solid glass plug has a threaded glass snelling which accepts an assembly similar to the one supplied with standard PTFE stopcocks. This consists of a PTFE washer, PTFE nut and Viton O-ring. Stopcock has a precision ground barrel and plug.
Catalog No. 50-153-1753 Supplier Chemglass Life Sciences Supplier No. CG45801
Solid glass plug has a threaded glass snelling which accepts an assembly similar to the one supplied with standard PTFE stopcocks. This consists of a PTFE washer, PTFE nut and Viton O-ring. Stopcock has a precision ground barrel and plug.