Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Glass Plug, Pressure, 4mm Bore, 17/40 Plug Size, 10mm OD Sidearms
SDP
Catalog No. CG45802
Encompass_Preferred
Solid glass plug has a threaded glass snelling which accepts an assembly similar to the one supplied with standard PTFE stopcocks. This consists of a PTFE washer, PTFE nut and Viton O-ring. Stopcock has a precision ground barrel and plug.
Catalog No. CG45802 Supplier Chemglass Life Sciences Supplier No. CG45802
Solid glass plug has a threaded glass snelling which accepts an assembly similar to the one supplied with standard PTFE stopcocks. This consists of a PTFE washer, PTFE nut and Viton O-ring. Stopcock has a precision ground barrel and plug.