Learn More
Chemglass Life Sciences Stopcock, 120° Bore, PTFE Plug, 4mm Bore, 16/35 Plug Size, 10mm OD Sidearms
SDP
Catalog No. CG44002
Encompass_Preferred
Stopcock, 120¿ Bore, PTFE Plug, 4mm Bore, 16/35 Plug Size, 10mm OD Sidearms. A 1:5 taper PTFE stopcock with three sidearms sealed 120¿ apart. Plug has a bore to allow connection of any two arms at once.
Catalog No. CG44002 Supplier Chemglass Life Sciences Supplier No. CG44002
Stopcock, 120¿ Bore, PTFE Plug, 4mm Bore, 16/35 Plug Size, 10mm OD Sidearms. A 1:5 taper PTFE stopcock with three sidearms sealed 120¿ apart. Plug has a bore to allow connection of any two arms at once.