Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Buret, PTFE Plug, 2mm Bore, 11/25 Plug Size, 8mm OD Sidearms, One Arm Is Pulled To A Delivery Tip
SDP
Catalog No. CG45201
Encompass_Preferred
Stopcock, Straight Bore, Buret, PTFE Plug, 2mm Bore, 11/25 Plug Size, 8mm OD Sidearms, One Arm Is Pulled To A Delivery Tip. For replacement or construction of burets having a 1:5 PTFE stopcock plug.
Catalog No. CG45201 Supplier Chemglass Life Sciences Supplier No. CG45201
Stopcock, Straight Bore, Buret, PTFE Plug, 2mm Bore, 11/25 Plug Size, 8mm OD Sidearms, One Arm Is Pulled To A Delivery Tip. For replacement or construction of burets having a 1:5 PTFE stopcock plug.