Learn More
Chemglass Life Sciences Stopcock, T-Bore, Glass Plug, Pressure, 2mm Bore, 17/40 Plug Size, 8mm OD Sidearms
SDP
Catalog No. CG45901
Encompass_Preferred
Similar to CG-458, but with three stems sealed 90º apart. Solid glass plug has a "T" shaped bore so that any two or all three arms can be connected at one time. Stopcock has a precision ground barrel and plug.
Catalog No. CG45901 Supplier Chemglass Life Sciences Supplier No. CG45901
Similar to CG-458, but with three stems sealed 90º apart. Solid glass plug has a "T" shaped bore so that any two or all three arms can be connected at one time. Stopcock has a precision ground barrel and plug.