Learn More
Chemglass Life Sciences Stopcock, T-Bore, Capillary Arms, PTFE Plug, 2mm Bore, 15.2/30 Plug Size, 7mm OD Sidearms
SDP
Catalog No. CG43801
Encompass_Preferred
Stopcock, T-Bore, Capillary Arms, PTFE Plug, 2mm Bore, 15.2/30 Plug Size, 7mm OD Sidearms. Similar to CG-436, but with all three arms being made of 2mm ID capillary tubing.
Catalog No. CG43801 Supplier Chemglass Life Sciences Supplier No. CG43801
Stopcock, T-Bore, Capillary Arms, PTFE Plug, 2mm Bore, 15.2/30 Plug Size, 7mm OD Sidearms. Similar to CG-436, but with all three arms being made of 2mm ID capillary tubing.