Learn More
Chemglass Life Sciences Stopcock, Double Oblique Bore, 1-Arm Capillary, PTFE Plug, 2mm Bore, 12.9/44 Plug Size, 7mm & 8mm OD Sidearms
SDP
Catalog No. 50873722
Encompass_Preferred
Stopcock, Double Oblique Bore, 1-Arm Capillary, PTFE Plug, 2mm Bore, 12.9/44 Plug Size, 7mm & 8mm OD Sidearms. Similar to CG-446, but with the top oblique stem being made of 2mm ID capillary tubing.
Catalog No. 50-873-722 Supplier Chemglass Life Sciences Supplier No. CG44801
Stopcock, Double Oblique Bore, 1-Arm Capillary, PTFE Plug, 2mm Bore, 12.9/44 Plug Size, 7mm & 8mm OD Sidearms. Similar to CG-446, but with the top oblique stem being made of 2mm ID capillary tubing.