Learn More
Chemglass Life Sciences Stopcock, T-Bore, PTFE Plug, 2mm Bore, 15.2/30 Plug Size, 8mm OD Sidearms
SDP
Catalog No. 50873718
Encompass_Preferred
Stopcock, T-Bore, PTFE Plug, 2mm Bore, 15.2/30 Plug Size, 8mm OD Sidearms. A 1:5 taper PTFE stopcock with three stems sealed 90º apart. PTFE plug has a "T" shaped bore to connect any two or all three stems.
Catalog No. 50-873-718 Supplier Chemglass Life Sciences Supplier No. CG43601
Stopcock, T-Bore, PTFE Plug, 2mm Bore, 15.2/30 Plug Size, 8mm OD Sidearms. A 1:5 taper PTFE stopcock with three stems sealed 90º apart. PTFE plug has a "T" shaped bore to connect any two or all three stems.