Learn More
Chemglass Life Sciences Stopcock, T-Bore, PTFE Plug, 4mm Bore, 16/35 Plug Size, 10mm OD Sidearms
SDP
Catalog No. 50873719
Encompass_Preferred
Stopcock, T-Bore, PTFE Plug, 4mm Bore, 16/35 Plug Size, 10mm OD Sidearms. A 1:5 taper PTFE stopcock with three stems sealed 90º apart. PTFE plug has a "T" shaped bore to connect any two or all three stems.
Catalog No. 50-873-719 Supplier Chemglass Life Sciences Supplier No. CG43602
Stopcock, T-Bore, PTFE Plug, 4mm Bore, 16/35 Plug Size, 10mm OD Sidearms. A 1:5 taper PTFE stopcock with three stems sealed 90º apart. PTFE plug has a "T" shaped bore to connect any two or all three stems.