Learn More
Med Vet International 9 Lives Autoclavable Surgical Skin Stapler & Cartridge of 35 Staples, Sterile, J1501H, Each
Supplier:  Med Vet International JOR1501H
Encompass_Preferred
Use every staple!!!! Fully autoclavable handle and cartridge Cartridge refills are autoclavable Cartridge refills are autoclavableHuman surgeons understand the value and use of skin staplers. Staplers are used in the vast majority of human surgeries. They now are also becoming popular in veterinary surgery. • faster closure to reduce surgery time • tolerated by patients, less chewing and licking the incision • now priced competitive against suture • simple technique with good cosmetic results Features: • Fixed angled head allows easy visualization during closure. • Simple to reload cartridges; for best results autoclave handle and cartridge separately. • Can view remaining staples on clear bottom. • Light weight but durable with comfortable ergonomic grip. • Handle measures 6.5" L x 3" H x 1/2" W • Staples are 316 L surgical stainless steel Cartridge measures 2 1/4" L x 3/4" W • Skin Staple after Closure: 3.6mm high x 6.9mm wide The Challenge
Catalog No. 50-004-95058