Learn More
Chemglass Life Sciences Stopper, Polyethylene, 24/25 and 24/40
SDP
Catalog No. CG302103
Encompass_Preferred
Stopper, Polyethylene, 24/25 and 24/40. Inexpensive, tapered stopper provides for a tight, leak-proof fit while the top ring permits the stopper to be easily removed. Stoppers are made of yellow Polyethylene. SOLD 100/PKG.
Catalog No. CG302103 Supplier Chemglass Life Sciences Supplier No. CG302103
Stopper, Polyethylene, 24/25 and 24/40. Inexpensive, tapered stopper provides for a tight, leak-proof fit while the top ring permits the stopper to be easily removed. Stoppers are made of yellow Polyethylene. SOLD 100/PKG.