Learn More
Halkey Roberts Corporation Straight male threaded swabable valve
SDP
Catalog No. NC1724653
Encompass
This item must be purchased in quantities of 2,000 or 8,000 EA per supplier requirements. Straight male threaded swabable valves utilize a female luer lock compatible connector and a male luer lock adapter. Mates securely with all standard luer syringes and connectors providing a hermetic seal between syringe luer tip and valve.
This item is not returnable. View return policy
Catalog No. NC1724653 Supplier Halkey Roberts Corporation Supplier No. 245204024
This item must be purchased in quantities of 2,000 or 8,000 EA per supplier requirements. Straight male threaded swabable valves utilize a female luer lock compatible connector and a male luer lock adapter. Mates securely with all standard luer syringes and connectors providing a hermetic seal between syringe luer tip and valve.