Learn More
Fine Science Tools Student Surgical Scissors, Stainless Steel, 14.5 cm
SDP
Catalog No. NC0127567
Encompass
Tip Shape: Straight/ Tips: Sharp-Blunt/ Cutting Edge: 45mm/ Alloy / Material: Student Stainless Steel/ Length: 14.5cm/ Serrated: No/ Feature: Student Quality
Catalog No. NC0127567 Supplier Fine Science Tools Supplier No. 9140114
Tip Shape: Straight/ Tips: Sharp-Blunt/ Cutting Edge: 45mm/ Alloy / Material: Student Stainless Steel/ Length: 14.5cm/ Serrated: No/ Feature: Student Quality