Learn More
MSC Eagle Lab Pack Open Head Drum, 55 Gal., HDPE, Blue, 1 ea.
Catalog No. 5000512192
Encompass
This 55 gal. open head drum is constructed of durable, blow-molded high-density polyethylene (HDPE) with UV inhibitors to provide excellent weather and chemical resistance. Featuring a white lid with a secure metal lever-lock band closure and two 2 in. bung holes, this straight-sided container is designed for the safe storage, handling, and transport of hazardous materials, bio-waste, and spill containment.
  • Constructed from high-density polyethylene for long-term durability
  • Equipped with UV inhibitors for improved environmental resistance
  • Includes a metal lever-lock band for a secure lid closure
  • Features two 2 in. bung holes for convenient access
  • Rated for hazardous material containment (UN 1H2/X200/S and 1H2/Y250/S)
  • Lightweight design allows for easier handling
Catalog No. 50-005-12192 Supplier MSC Supplier No. 54449392
This 55 gal. open head drum is constructed of durable, blow-molded high-density polyethylene (HDPE) with UV inhibitors to provide excellent weather and chemical resistance. Featuring a white lid with a secure metal lever-lock band closure and two 2 in. bung holes, this straight-sided container is designed for the safe storage, handling, and transport of hazardous materials, bio-waste, and spill containment.
  • Constructed from high-density polyethylene for long-term durability
  • Equipped with UV inhibitors for improved environmental resistance
  • Includes a metal lever-lock band for a secure lid closure
  • Features two 2 in. bung holes for convenient access
  • Rated for hazardous material containment (UN 1H2/X200/S and 1H2/Y250/S)
  • Lightweight design allows for easier handling