Learn More
Andwin Scientific STR TIPS 96 PER RACK
SDP
Catalog No. NC9195754
Encompass_Preferred
The Extra Volume Pipet Tips by International is made to simplify aspirating from and dispensing into PCR tubes and plates. The tips are made for Matrix Impact2™ Pipettors. These are constructed from polypropylene, are gamma-irradiated, and are autocla
Catalog No. NC9195754 Supplier Andwin Scientific Supplier No. 53508920
The Extra Volume Pipet Tips by International is made to simplify aspirating from and dispensing into PCR tubes and plates. The tips are made for Matrix Impact2™ Pipettors. These are constructed from polypropylene, are gamma-irradiated, and are autocla