Learn More
Chemglass Life Sciences Stir Vanes, PTFE, 5.25mm Width X 11mm Height
SDP
Catalog No. 50872790
Encompass_Preferred
Stir Vanes, PTFE, 5.25mm Width X 11mm Height. Stir Bars, PTFE Covered, for Microwave vials. Manufactured with FDA-grade and USP Class IV PTFE.
Catalog No. 50-872-790 Supplier Chemglass Life Sciences Supplier No. CG492017
Stir Vanes, PTFE, 5.25mm Width X 11mm Height. Stir Bars, PTFE Covered, for Microwave vials. Manufactured with FDA-grade and USP Class IV PTFE.