Learn More
Med Vet International Miltex Vantage Strabismus Scissors, 4in, Curved
Catalog No. 502643474
Encompass_Preferred
The Miltex Vantage V95-314 Strabismus Scissors are made of mid-grade stainless steel and has a length of 4in. This Ophthalmic dissecting instrument features round blunt blades and a curved shank. The Miltex Vantage Strabismus Scissors are commmonly used in procedures where delicate maneuvering is required.
Catalog No. 50-264-3474 Supplier Med Vet International Supplier No. MILV95314
The Miltex Vantage V95-314 Strabismus Scissors are made of mid-grade stainless steel and has a length of 4in. This Ophthalmic dissecting instrument features round blunt blades and a curved shank. The Miltex Vantage Strabismus Scissors are commmonly used in procedures where delicate maneuvering is required.