Learn More
Med Vet International Miltex Vantage Strabismus Scissors, 4in., Straight
Catalog No. 502830576
Encompass_Preferred
The Miltex Vantage V95-312 Strabismus Scissors are made of mid-grade stainless steel and has a length of 4in. This Ophthalmic dissecting instrument features round blunt blades and a straight shank. The Miltex Vantage Strabismus Scissors are commonly used in procedures where delicate maneuvering is required.
Catalog No. 50-283-0576 Supplier Med Vet International Supplier No. MILV95312
The Miltex Vantage V95-312 Strabismus Scissors are made of mid-grade stainless steel and has a length of 4in. This Ophthalmic dissecting instrument features round blunt blades and a straight shank. The Miltex Vantage Strabismus Scissors are commonly used in procedures where delicate maneuvering is required.