Learn More
Fine Science Tools STRABISMUS SCISSORS
SDP
Catalog No. NC9266917
Encompass
Tip Shape: Curved/ Tips: Blunt-Blunt/ Cutting Edge: 23mm/ Tip Diameter: / Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No
Catalog No. NC9266917 Supplier Fine Science Tools Supplier No. 1407511
Tip Shape: Curved/ Tips: Blunt-Blunt/ Cutting Edge: 23mm/ Tip Diameter: / Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No