Learn More
Fine Science Tools Strabismus Scissors, Stainless Steel, 9 cm
SDP
Catalog No. NC9367633
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 19mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: No
Catalog No. NC9367633 Supplier Fine Science Tools Supplier No. 1407409
Add to Cart
Add to Cart
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 19mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: No