Learn More
Alkali Scientific Storage Box, Hinged Lid, Rainbow, 100 x 1.5mL, 5/cs
SDP
Catalog No. 502336032
Encompass_Preferred
ASI's Suitable for refrigerator and freezer storage as low as -80˚C, Tube Storage Box have translucent Lids and numbered cavities. Polypropylene, withstand -80˚C. 6 types - for 0.2ml to 50ml tubes. Compact and space efficient. Autoclavable with numbered tube wells. Rainbow pack of five colors.
Catalog No. 50-233-6032 Supplier Alkali Scientific Supplier No. FS100A
ASI's Suitable for refrigerator and freezer storage as low as -80˚C, Tube Storage Box have translucent Lids and numbered cavities. Polypropylene, withstand -80˚C. 6 types - for 0.2ml to 50ml tubes. Compact and space efficient. Autoclavable with numbered tube wells. Rainbow pack of five colors.