Learn More
Fine Science Tools Fine Scissors, Extra Large Loops, Stainless Steel, 9.5 cm
SDP
Catalog No. NC9007345
Encompass
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 14mm/ Alloy / Material: Stainless Steel/ Length: 9.5cm/ Serrated: No/ Feature: Large Loops
Catalog No. NC9007345 Supplier Fine Science Tools Supplier No. 1410609
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 14mm/ Alloy / Material: Stainless Steel/ Length: 9.5cm/ Serrated: No/ Feature: Large Loops