Learn More
MSC Norgren Straight Adapter, 1/4 in OD Tube, NPTF, 1 ea
Catalog No. 501920143
Encompass
This straight adapter is a push-to-connect fitting designed for reliable connections in pneumatic systems. It facilitates the connection of a 1/4 in OD tube to an MNPT fitting.
  • Brass construction
  • Pressure rating of 150 psi
  • Nitrile O-ring material
  • Suitable for general purpose use
  • Temperature range of 0–175 °F
Catalog No. 50-192-0143 Supplier MSC Supplier No. 00348912
This straight adapter is a push-to-connect fitting designed for reliable connections in pneumatic systems. It facilitates the connection of a 1/4 in OD tube to an MNPT fitting.
  • Brass construction
  • Pressure rating of 150 psi
  • Nitrile O-ring material
  • Suitable for general purpose use
  • Temperature range of 0–175 °F