Learn More
Bellco Glass, Inc. Straight Adapter,1/4x1/8" Barbed x Male pipe
SDP
Supplier:  Bellco Glass, Inc. 780800005
Encompass_Preferred
Straight tube to pipe adapter. MADE IN THE USABarbed on one end & male pipe on other endFor tube id: 1/4"Pipe size: 1/8"nptMaterial: white polypropyleneMax. Pressure: 125psi @72°fTemp. Range: -40° to +200°fUse with air & waterMaterial meets: ANSI/HIMA md 70.1 & iso 594/1-1986Tubing: use PVC, polyurethane, and silicone tubing with durometer of- A75-A85 Clamps: not requiredCASE/PKG: 10
Catalog No. 50-194-6384