Learn More
Glass Expansion Thermo Pro Glass Straight Ball Joint Adapter
SDP
Catalog No. 502703179
Encompass_Preferred
Glass straight ball joint adaptor compatible with Thermo 6000/7000 Radial Series, Pro Radial and Pro Duo.
Catalog No. 50-270-3179 Supplier Glass Expansion Supplier No. 218092723
Glass straight ball joint adaptor compatible with Thermo 6000/7000 Radial Series, Pro Radial and Pro Duo.