Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampco Straight Blade Hoe with Fiberglass Handle
SDP

Catalog No. 19173925 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English) Blade:
10 in.
11 -1/2 in.
6 in.
8 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Blade
19-173-925 11 -1/2 in.
19-173-922 6 in.
19-173-923 8 in.
19-173-924 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-173-925 Supplier Ampco Supplier No. H103FG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Straight blade hoe for garden and mixing purposes.

  • Handle included
  • Often purchased with Ampco rake, hand pick, or shovel.

Specifications

Product Type Garden Hoe
Length (English) 55 in.
Length (English) Blade 11 -1/2 in.
Quantity 1 Ea.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.