Learn More
Chemglass Life Sciences Stopcock, Straight Bore, PTFE Metering Plug, 2mm Bore, 11/25 Plug Size, 8mm OD Sidearms
SDP
Catalog No. CG43401
Encompass_Preferred
Stopcock, Straight Bore, PTFE Metering Plug, 2mm Bore, 11/25 Plug Size, 8mm OD Sidearms. A 1:5 taper PTFE plug having a threaded needle valve to allow the adjustment of flow through the bore of the plug.
Catalog No. CG43401 Supplier Chemglass Life Sciences Supplier No. CG43401
Stopcock, Straight Bore, PTFE Metering Plug, 2mm Bore, 11/25 Plug Size, 8mm OD Sidearms. A 1:5 taper PTFE plug having a threaded needle valve to allow the adjustment of flow through the bore of the plug.