Learn More
Honeywell™ PA700 PAPR System Straight Breathing Tube
Catalog No. 19085509
Change view
Click to view available options
Length (English):
34 in.
Length (Metric):
86.3 cm
Size:
Regular
Catalog No. Length (English) Length (Metric) Size
19-085-509 34 in. 86.3 cm Regular
1 options
This item is not returnable. View return policy
Catalog No. 19-085-509 Supplier Honeywell™ Supplier No. PA031
This item is not returnable. View return policy
Breathing tube designed for use with PA 700 Series Powered Air Purifying Respirator (PAPR).
  • Tube features simple, twist and lock connectors
  • Lightweight, highly flexible corrugated plastic hose material for freedom of movement
  • Swivel connection prevents hose from twisting or binding
  • Continuous flow components
  • Straight
TRUSTED_SUSTAINABILITY
Specifications
Size Regular
For Use With (Equipment) Primair 700 Series PAPR
Model PA031
Length (English) 34 in.
Length (Metric) 86.3 cm