Learn More
Fine Science Tools Dietrich Bulldog Clamps, Stainless Steel, 52.5 mm
SDP
Catalog No. NC1571805
Encompass
Product: Dietrich Bulldog Clamp - Straight/52.5mm/ Shape: Straight/ Jaw Dimensions: 8.6 x 1.3mm/ Clamping Pressure: Light/ Gram Pressure: 50g/ Alloy: Stainless Steel/ Length: 52.5mm/ Serrated: Yes/ Unit: Each
This item is not returnable. View return policy
Catalog No. NC1571805 Supplier Fine Science Tools Supplier No. 1803845
Product: Dietrich Bulldog Clamp - Straight/52.5mm/ Shape: Straight/ Jaw Dimensions: 8.6 x 1.3mm/ Clamping Pressure: Light/ Gram Pressure: 50g/ Alloy: Stainless Steel/ Length: 52.5mm/ Serrated: Yes/ Unit: Each