Learn More
Chemglass Life Sciences 30" Straight Connector with Male Luer Lock, L/S Platinum Cured Silicone Pump Tubing, 1/8" ID x 1/4" OD Size 16, PK10
SDP
Catalog No. 5000083130
Encompass
Reduces set up time and minimizes labor cost. straight connector and male luer assembly are package 10 per bag and ready to use.Manufactured with 30' of No.18 (1/8 x 1/4') platinum cured silicone tubing.For sterile applications the assembly can be safely autoclaved.
Catalog No. 50-000-83130 Supplier Chemglass Life Sciences Supplier No. CLS139023
Reduces set up time and minimizes labor cost. straight connector and male luer assembly are package 10 per bag and ready to use.Manufactured with 30' of No.18 (1/8 x 1/4') platinum cured silicone tubing.For sterile applications the assembly can be safely autoclaved.