Learn More
Research Products International Corp Straight Tube Connectors for 3/8 Inch Tubing, 12 per Bag
SDP
Supplier:  Research Products International Corp F19509
Encompass
These straight fittings ensure a tight, secure connection between lengths of tubing having the same inner diameter (I.D.). Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases. They are FDA-grade for direct contact with food, drugs, and cosmetics. Autoclavable.
Catalog No. 50-998-408