Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight, Hose Barb Union, 3/32"" ID; 25/PK
Catalog No. NC9560166
Encompass
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. NC9560166 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0636533
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.