Learn More
Accuflow Systems Masterflex Hose Barb Reducer, PVDF, 1/4 in to 1/8 in
Catalog No. NC2905576
Encompass_Preferred
Straight PVDF hose barb reducer for connecting 1/4 in ID tubing to 1/8 in ID tubing. Chemically resistant PVDF construction provides dimensional stability and compatibility with common laboratory solvents; suitable for low-pressure fluid routing and single-use assemblies.
  • Provides secure push-fit connection between 1/4 in and 1/8 in ID tubing.
  • Chemical resistance to a wide range of solvents and acids.
  • Suitable for low-pressure and single-use applications.
  • Rigid PVDF construction for dimensional stability.
  • Typically packaged in 10-piece sets for bulk laboratory use.
Catalog No. NC2905576 Supplier Accuflow Systems Supplier No. 4061456
Straight PVDF hose barb reducer for connecting 1/4 in ID tubing to 1/8 in ID tubing. Chemically resistant PVDF construction provides dimensional stability and compatibility with common laboratory solvents; suitable for low-pressure fluid routing and single-use assemblies.
  • Provides secure push-fit connection between 1/4 in and 1/8 in ID tubing.
  • Chemical resistance to a wide range of solvents and acids.
  • Suitable for low-pressure and single-use applications.
  • Rigid PVDF construction for dimensional stability.
  • Typically packaged in 10-piece sets for bulk laboratory use.