Learn More
Grainger Straight Cut Profile Router Bit 1/4
Catalog No. 502765549
Encompass_Preferred
The WESTWARD Straight Cut Profile Router is a handheld power tool designed for shaping and trimming wood, plastic, and soft metals. It has a 1-1/4 HP motor with a variable speed range of 10,000-30,000 RPM. The router features a micro-adjustable depth stop, a soft grip handle, and a quick release cam lock system for easy bit changes. It comes with a 1/4" collet, chip deflector, wrenches, and an operator's manual. This versatile router is ideal for trimming laminates and veneers, cutting dadoes, rabbets, and grooves.
Catalog No. 50-276-5549 Supplier Grainger Supplier No. 16Y634
The WESTWARD Straight Cut Profile Router is a handheld power tool designed for shaping and trimming wood, plastic, and soft metals. It has a 1-1/4 HP motor with a variable speed range of 10,000-30,000 RPM. The router features a micro-adjustable depth stop, a soft grip handle, and a quick release cam lock system for easy bit changes. It comes with a 1/4" collet, chip deflector, wrenches, and an operator's manual. This versatile router is ideal for trimming laminates and veneers, cutting dadoes, rabbets, and grooves.