Learn More
Grainger Straight Cut Profile Router Bit 1/4
Catalog No. 502767824
Encompass_Preferred
The Bosch Fractional Router Bit Set contains 8 router bits made of high-speed steel. The set includes 1/4", 3/8", 1/2" straight bits, 1/4" and 1/2" roundover bits, 1/4" and 1/2" cove bits, and a 1/4" chamfer bit. These bits are designed for use in handheld and stationary routers and are ideal for decorative edging, profiling, hinge mortising, slot cutting, and other woodworking applications. The bits feature precision ground carbide tips for long life and clean cuts in wood, plastic and soft metals. The 1/4" shanks fit most common router collets. This versatile 8-piece router bit set provides a useful assortment of profiles for woodworking projects.
Catalog No. 50-276-7824 Supplier Grainger Supplier No. 16Y583
The Bosch Fractional Router Bit Set contains 8 router bits made of high-speed steel. The set includes 1/4", 3/8", 1/2" straight bits, 1/4" and 1/2" roundover bits, 1/4" and 1/2" cove bits, and a 1/4" chamfer bit. These bits are designed for use in handheld and stationary routers and are ideal for decorative edging, profiling, hinge mortising, slot cutting, and other woodworking applications. The bits feature precision ground carbide tips for long life and clean cuts in wood, plastic and soft metals. The 1/4" shanks fit most common router collets. This versatile 8-piece router bit set provides a useful assortment of profiles for woodworking projects.