Learn More
Grainger Straight Cut Profile Router Bit 3/4
Catalog No. 502766271
Encompass_Preferred
The Westward Straight Router Bit with Carbide Tipped 1/4" Shank is a high speed steel bit with a carbide tip for long life. It has a straight cutting profile and a 1/4" shank diameter. The overall length is 3-1/2" and it can cut grooves up to 1/4" wide and 1/2" deep in wood, plastic and soft metals. The carbide tipped design allows for smooth, fast cuts without overheating. This versatile bit is good for general purpose routing work.
Catalog No. 50-276-6271 Supplier Grainger Supplier No. 16Y581
The Westward Straight Router Bit with Carbide Tipped 1/4" Shank is a high speed steel bit with a carbide tip for long life. It has a straight cutting profile and a 1/4" shank diameter. The overall length is 3-1/2" and it can cut grooves up to 1/4" wide and 1/2" deep in wood, plastic and soft metals. The carbide tipped design allows for smooth, fast cuts without overheating. This versatile bit is good for general purpose routing work.