Learn More
Grainger Straight Cut Profile Router Bit 1/2
Catalog No. 502766898
Encompass_Preferred
The WESTWARD Straight Cut Profile Router is a handheld power tool for shaping and trimming wood, plastic, and soft metals. It has a 1-1/4 HP motor with a variable speed range of 10,000-30,000 RPM. The router accepts 1/4" and 1/2" shank cutters up to 1-1/2" in diameter. It features a soft start, electronic speed control, and a micro-depth adjustment system for precise depth adjustments in 1/128" increments. It includes a fixed base, 1/4" and 1/2" collets, chip deflector, and carrying case. Ideal for trimming laminates, cutting dadoes, rabbeting edges, and other edge forming applications.
Catalog No. 50-276-6898 Supplier Grainger Supplier No. 16Y586
The WESTWARD Straight Cut Profile Router is a handheld power tool for shaping and trimming wood, plastic, and soft metals. It has a 1-1/4 HP motor with a variable speed range of 10,000-30,000 RPM. The router accepts 1/4" and 1/2" shank cutters up to 1-1/2" in diameter. It features a soft start, electronic speed control, and a micro-depth adjustment system for precise depth adjustments in 1/128" increments. It includes a fixed base, 1/4" and 1/2" collets, chip deflector, and carrying case. Ideal for trimming laminates, cutting dadoes, rabbeting edges, and other edge forming applications.