Learn More
Grainger Straight Cut Profile Router Bit 3/16
Catalog No. 502765076
Encompass_Preferred
The Westward Straight Cut Profile Router Bit is a 3/16" diameter high-speed steel bit with a 1/4" shank. It has a straight cutting profile and is designed for use in handheld or table-mounted routers. The bit features a TiN coating for increased durability and smooth cutting action. With its straight profile, this bit is well-suited for cutting grooves, slots and edges in wood, plastic and soft metals. The bit has an overall length of 1-1/2" and comes with a plastic case for convenient storage when not in use. This is a high-quality American-made router bit good for precision cutting applications.
Catalog No. 50-276-5076 Supplier Grainger Supplier No. 16Y596
The Westward Straight Cut Profile Router Bit is a 3/16" diameter high-speed steel bit with a 1/4" shank. It has a straight cutting profile and is designed for use in handheld or table-mounted routers. The bit features a TiN coating for increased durability and smooth cutting action. With its straight profile, this bit is well-suited for cutting grooves, slots and edges in wood, plastic and soft metals. The bit has an overall length of 1-1/2" and comes with a plastic case for convenient storage when not in use. This is a high-quality American-made router bit good for precision cutting applications.