Learn More
Grainger Straight Cut Profile Router Bit 1/2
Catalog No. 502765224
Encompass_Preferred
The WESTWARD Straight Cut Profile Router is a handheld power tool designed for shaping and trimming wood, plastic, and soft metals. It has a 1-1/4 HP motor with a variable speed range of 10,000-30,000 RPM. The router features a micro-adjustable depth stop, a soft grip handle, and a quick release cam lock system for easy bit changes. It comes with a 1/4" and 1/2" collet, chip deflector, straight guide fence, and plastic case. This versatile router is ideal for trimming laminates and edging profiles.
Catalog No. 50-276-5224 Supplier Grainger Supplier No. 16Y573
The WESTWARD Straight Cut Profile Router is a handheld power tool designed for shaping and trimming wood, plastic, and soft metals. It has a 1-1/4 HP motor with a variable speed range of 10,000-30,000 RPM. The router features a micro-adjustable depth stop, a soft grip handle, and a quick release cam lock system for easy bit changes. It comes with a 1/4" and 1/2" collet, chip deflector, straight guide fence, and plastic case. This versatile router is ideal for trimming laminates and edging profiles.