Learn More
Fine Science Tools Dowell Scissors, Straight, 11.5 cm
SDP
Catalog No. NC1423879
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 10mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No
Catalog No. NC1423879 Supplier Fine Science Tools Supplier No. 1504011
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 10mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No